Learn More
Cayman Chemical ANTIBODY ML-141 1mg
SDP
Supplier:  Cayman Chemical 184961 MG
Encompass_Preferred
A noncompetitive, allosteric inhibitor of Cdc42 (EC50 = 2.1 µM) with no inhibitory action against Rho, Ras, Rab, or Rac; also inhibits the constitutively active Cdc42 Q61L mutant (EC50 = 2.6 µM)
Catalog No. 50-298-0451