Learn More
Cayman Chemical ANTIBODY ML-192 10mg
SDP
Supplier:  Cayman Chemical 1518310 MG
Encompass_Preferred
A selective GPR55 antagonist (IC50 = 702 nM); selective for GPR55 over CB1 and CB2, as well as GPR35 (IC50s = >32 μM)
Catalog No. 50-297-0953