Learn More
Cayman Chemical ANTIBODY ML-198 1mg
SDP
Supplier:  Cayman Chemical 377551 MG
Encompass_Preferred
A chemical chaperone of glucocerebrosidase; activates hydrolysis of 4-MUG in spleen homogenate isolated from patients with Gaucher disease with the homozygous glucocerebrosidaseN370S mutation (AC50 = 0.36 µM); increases the translocation of glucocerebrosidaseN370S to lysosomes in dermal fibroblasts isolated from patients with Gaucher disease at 5 µM
Catalog No. 50-291-6861