Learn More
Cayman Chemical ANTIBODY ML-202 25mg
SDP
Supplier:  Cayman Chemical 3732625 MG
Encompass_Preferred
A PKM2 activator; induces PMK2 activation in a cell-free assay (AC50 = 73 nM)
Catalog No. 50-291-9296