Learn More
Cayman Chemical ANTIBODY ML-204 50mg
SDP
Supplier:  Cayman Chemical 1562650 MG
Encompass_Preferred
Selectively blocks TRPC4 channels (IC50s = 0.96 and 2.6 μM in fluorescent and electrophysiological assays, respectively)
Catalog No. 50-297-2654