Learn More
Cayman Chemical ANTIBODY ML-221 10mg
SDP
Supplier:  Cayman Chemical 2731310 MG
Encompass_Preferred
An APJ receptor antagonist (IC50 = 4.8 μM); selective for APJ over AT1 (IC50 = >78 μM); antagonizes apelin 13-induced activation of APJ in cAMP and β-arrestin recruitment assays (IC50s = 0.7 and 1.75 μM, respectively); inhibits proliferation and angiogenesis in Mz-ChA-1 cholangiocarcinoma cells from 5-15 μM; reduces tumor growth in a Mz-ChA-1 mouse xenograft model at 150 μg/kg; reduces sciatic CCI-induced mechanical allodynia and heat hyperalgesia in rats at 10 μg/animal; inhibits pathological angiogenesis and enhances normal vessel recovery in retinal ischemic regions in a mouse model of oxygen-induced retinopathy
Catalog No. 50-293-8578