Learn More
Cayman Chemical ANTIBODY ML-226 5mg
SDP
Supplier:  Cayman Chemical 256815 MG
Encompass_Preferred
An inhibitor of ABHD11 (IC50s = 15 and 0.68 nM, respectively, in vitro and in situ); 100-fold selective for ABHD11 over approximately 20 serine hydrolases; can be used as an anti-probe for ML-211
Catalog No. 50-293-3116