Learn More
Cayman Chemical ANTIBODY ML-228 50mg
SDP
Supplier:  Cayman Chemical 1456850 MG
Encompass_Preferred
An activator of the HIF signaling pathway, as demonstrated by HRE reporter assay (EC50 = 1.2 µM), HIF-1α nuclear translocation assay (EC50 = 1.3 µM), and increased VEGF expression (EC50 = 1.6 µM); activity is blocked by excess iron, suggesting that ML-228 can chelate iron
Catalog No. 50-297-0926