Learn More
Cayman Chemical ANTIBODY ML-334 10mg
SDP
Supplier:  Cayman Chemical 3143910 MG
Encompass_Preferred
An inhibitor of the Nrf2-Keap1 protein-protein interaction; binds to the Keap1 Kelch domain (IC50 = 1 µM) to induce dissociation and nuclear translocation of Nrf2 and activation of the ARE in a cell-based assay (EC50 = 12 µM)
Catalog No. 50-299-2093