Learn More
Cayman Chemical ANTIBODY ML-347 10mg
SDP
Supplier:  Cayman Chemical 1764010 MG
Encompass_Preferred
Selectively inhibits ALK1 (IC50 = 46 nM) and ALK2 (IC50 = 32 nM) with >300-fold selectivity over ALK3, ALK6, and VEGF type 2 receptor; inhibits BMP4 signaling with an IC50 value of 152 nM in a functional assay
Catalog No. 50-298-0167