Learn More
Cayman Chemical ANTIBODY ML-349 1mg
SDP
Supplier:  Cayman Chemical 209231 MG
Encompass_Preferred
A selective and reversible inhibitor of LYPLA2 (Kis = 230 and >10,000 nM for human recombinant LYPLA2 and LYPLA1, respectively); inhibits LYPLA2 (>95%) with no change in the activity of 25 other serine hydrolases in BW5147 murine T cell hybridoma cells; almost completely inhibits LYPLA2 in HEK293T and mouse T cells and in vivo in murine lung, heart, and kidneys; partially inhibits LYPLA2 in murine brain
Catalog No. 50-298-3975