Learn More
Cayman Chemical ANTIBODY ML-354 5mg
SDP
Supplier:  Cayman Chemical 191205 MG
Encompass_Preferred
A PAR4 antagonist (IC50 = 140 nM) that exhibits 71-fold selectivity for PAR4 over PAR1 (IC50 = 10 μM) in isolated human platelets
Catalog No. 50-298-1207