Learn More
Cayman Chemical ANTIBODY ML-396 1mg
SDP
Supplier:  Cayman Chemical 291671 MG
Encompass_Preferred
A positive allosteric modulator of group III mGluRs (EC50s = 108, 146, and 128 nM for mGluR4, mGluR7, and mGluR8, respectively, in calcium mobilization assays); potentiates LSP4-2022-induced reductions in fEPSP slopes in mouse hippocampal slices; restores hippocampal long-term potentiation, improves novel object recognition, contextual fear learning, and social memory, and decreases sleep apneas in the Mecp2-/- mouse model of Rett syndrome at 30 mg/kg
Catalog No. 50-293-9985