Learn More
Cayman Chemical ANTIBODY MLE-4901 50mg
SDP
Supplier:  Cayman Chemical 2358750 MG
Encompass_Preferred
An NK3 receptor antagonist; inhibits human liver microsomal CYP3A4/5 (apparent IC50s = 7.1 and 19.8 μM in midazolam 1'-hydroxylation and testosterone 6β-hydroxylation assays, respectively)
Catalog No. 50-299-4237