Learn More
Cayman Chemical ANTIBODY MLS-573151 25mg
SDP
Supplier:  Cayman Chemical 1526925 MG
Encompass_Preferred
A specific, cell permeable inhibitor of Cdc42 (IC50 = 2 μM); has no effect on other GTPases, including the related Rho family members RhoA, Rac1, and Rac2
Catalog No. 50-296-8530