Learn More
Cayman Chemical ANTIBODY Monastrol 5mg
SDP
Supplier:  Cayman Chemical 150445 MG
Encompass_Preferred
A reversible, cell-permeable inhibitor of the motor protein Eg5, blocking basal ATPase activity in vitro (IC50 = 6.1 µM) and inducing the formation of monoastral spindles in cell-based assays (IC50 = 51.3 μM)
Catalog No. 50-296-9884