Learn More
Sino Biological Recombinant Anti-IL-13 Antibody, Rabbit Monoclonal 100 µL
Supplier:  Sino Biological 50225R062100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse IL-13 (rM IL-13; Catalog#50225-MNAH; P20109; Met1-Phe131).
Catalog No. 50-007-49842