Learn More
Cayman Chemical ANTIBODY Mps1-IN-1 500ug
SDP
Supplier:  Cayman Chemical 19175500 UG
Encompass_Preferred
A selective inhibitor of Mps1 kinase (IC50 = 367 nM); disrupts the recruitment of Mad2 to kinetochores and increases the frequency of multipolar mitosis in U2OS cells
Catalog No. 50-298-4610