Learn More
Cayman Chemical ANTIBODY MRK-740 5mg
SDP
Supplier:  Cayman Chemical 263875 MG
Encompass_Preferred
A PRDM9 inhibitor (IC50 = 80 nM); inhibits PRDM9-dependent trimethylation of ectopic histone H3K4 (IC50 = 0.8 µM) and endogenous H3K4 in HEK293 cells in a concentration-dependent manner; induces meiotic defects in mouse spermatocytes
Catalog No. 50-293-3757