Learn More
Cayman Chemical ANTIBODY MRS1754 25mg
SDP
Supplier:  Cayman Chemical 3350125 MG
Encompass_Preferred
An adenosine A2B receptor antagonist (Ki = 1.97 nM); selective for adenosine A2B receptors over adenosine A1, A2A, and A3 receptors (Kis = 403, 503, and 570 nM, respectively); increases 28-day survival, as well as decreases peritoneal bacterial growth and plasma levels of IL-6, TNF-α, and MIP-2, in a mouse model of CLP-induced sepsis from 0.5-10 mg/kg; reduces disease severity in a mouse model of EAE at 1 mg/kg
Catalog No. 50-294-7794