Learn More
Cayman Chemical ANTIBODY MS023 10mg
SDP
Supplier:  Cayman Chemical 3478610 MG
Encompass_Preferred
An inhibitor of type I PRMTs (IC50s = 30, 119, 83, 4, and 5 nM for PRMT1, -3, -4, -6, and -8, respectively); selective for these type I PRMTs over PRMT5, -7, and -9, as well as a panel of 25 PKMTs and DNMTs at 10 µM; inhibits methylation of H4R3 in MCF-7 cells and H3R2 in HEK293 cells (IC50s = 9 and 56 nM, respectively); inhibits methylation of the SARS-CoV-2 nucleocapsid protein and the replication of SARS-CoV-2 in Vero E6 cells at 20 µM; reduces tumor growth in a Huh7 mouse xenograft model at 160 mg/kg
Catalog No. 50-291-2663