Learn More
Cayman Chemical ANTIBODY-mT5-14 5mg
SDP
Supplier:  Cayman Chemical 378295 MG
Encompass_Preferred
A derivative of remdesivir with antiviral activity; increases the viability of Vero E6 cells infected with SARS-CoV-2 (EC50 = 0.4 µM), as well as Vero E6 cells infected with the Alpha, Beta, Gamma, or Delta SARS-CoV-2 variants (EC50s = 2.5, 15.9, 1.7, and 5.6 µM, respectively); exhibits increased plasma and lung concentrations compared with remdesivir in hamsters at 10 mg/kg
Catalog No. 50-291-5547