Learn More
Cayman Chemical ANTIBODY MW-150 5mg
SDP
Supplier:  Cayman Chemical 337895 MG
Encompass_Preferred
An inhibitor of p38α MAPK (Ki = 101 nM); 6-, 10-, and 14-fold selective for p38α MAPK over NLK, p38β MAPK, and p38δ MAPK, respectively; selective for p38α MAPK over p38α MAPKT106M; inhibits p38α MAPK phosphorylation of MK2 in LPS-activated glia; improves spatial reference memory in the radial arm water maze in aged APP/PS1 mice
Catalog No. 50-294-8707