Learn More
Cayman Chemical ANTIBODY MXS 5mg
SDP
Supplier:  Cayman Chemical 327265 MG
Encompass_Preferred
A fluorescent probe for hypochlorous acid; selectively fluoresces in the presence of OCl- over O2, H2O2, NO, NO2-, NO3-, O2-, ONOO-, and •OH at 10 µM; has been used to monitor real-time mitochondrial HOCl levels in living cells; excitation/emission maxima = 530/654 nm, respectively
Catalog No. 50-294-8591