Learn More
Cayman Chemical ANTIBODY Myr-ZIP 10mg
SDP
Supplier:  Cayman Chemical 3444510 MG
Encompass_Preferred
A PKMζ inhibitor (Ki = 0.076-2.11 µM); inhibits PKCα at 10 µM; prevents ionomycin-induced PKC translocation to the plasma membrane in HEK293 cells expressing AKAP79 at 5 µM; inhibits the conditioned place preference response to morphine in rats when administered intracranially into the nucleus accumbens core at doses of 10 and 30 nmol/0.5 µl per side
Catalog No. 50-294-8519