Learn More
Cayman Chemical ANTIBODY n-CHMIMO 1mg
SDP
Supplier:  Cayman Chemical 293011 MG
Encompass_Preferred
An analytical reference standard categorized as a synthetic cannabinoid; has been detected in human hair samples tested for verification of drug abstinence; intended for research and forensic applications
Catalog No. 50-298-8919