Learn More
Cayman Chemical ANTIBODY N106 5mg
SDP
Supplier:  Cayman Chemical 309525 MG
Encompass_Preferred
An activator of SERCA2A SUMOylation; activates SUMO-activating enzyme E1, which increases SUMOylation and the activity of SERCA2A in isolated adult rat cardiomyocytes from 0.1-1 µM; increases contractility in the same cells; increases cardiac SERCA2A SUMOylation and contractility in a mouse model of pressure overload-induced heart failure at 10 mg/kg
Catalog No. 50-298-9799