Learn More
Cayman Chemical ANTIBODY NAB 2 10mg
SDP
Supplier:  Cayman Chemical 2614510 MG
Encompass_Preferred
An inhibitor of α-synuclein-induced toxicity; prevents α-synuclein-induced defects in Rsp5-dependent endocytosis and vacuolar delivery of Mup1, Golgi-to-vacuole trafficking of Sna3, and degradation of Bap2 in wild-type yeast cells; prevents formation of α-synuclein foci in wild-type yeast cells
Catalog No. 50-293-6749