Learn More
Cayman Chemical ANTIBODY namorelIn 50mg
SDP
Supplier:  Cayman Chemical 2467650 MG
Encompass_Preferred
A small molecule agonist of GHS-R1a (Ki = 0.7 nM); induces calcium mobilization in CHO cells expressing rat GHS-R1a in a concentration-dependent manner and GH release from rat pituitary cells (EC50 = 1.5 nM); stimulates GH release in sham and vagotomized rats; increases food intake in rats
Catalog No. 50-293-5775