Learn More
Cayman Chemical ANTIBODY NCT-502 1mg
SDP
Supplier:  Cayman Chemical 197161 MG
Encompass_Preferred
A selective PHGDH inhibitor that blocks serine synthesis from 3-phosphoglycerate in cells (IC50 = 3.7 µM); reversibly blocks the production of glucose-derived serine in cells
Catalog No. 50-298-2838