Learn More
Cayman Chemical ANTIBODY nglIforIb 5mg
SDP
Supplier:  Cayman Chemical 384195 MG
Encompass_Preferred
A glycogen phosphorylase inhibitor (IC50s = 52, 352, and 150 nM for the liver, muscle, and brain form, respectively); decreases myocardial glycogen phosphorylase activity and infarct size as a percentage of the area at risk in a rabbit model of coronary artery occlusion-induced ischemia-reperfusion injury
Catalog No. 50-291-8421