Learn More
Cayman Chemical ANTIBODY NI-42 25mg
SDP
Supplier:  Cayman Chemical 2829325 MG
Encompass_Preferred
An inhibitor of the BRPF1 bromodomain (IC50 = 7.9 nM); selective for BRPF1 over BRPF2/BRD1, BRPF3, BRD9, and BRD4/BD1 (IC50s = 48, 260, 310, and 4,500 nM, respectively); inhibits the growth of OCI-AML2, Nomo-1, THP-1, KG-1, and MV-4-11 AML cells (GI50s = 1.3, 4.6, 5.7, 7, and 9.9 µM, respectively) and a variety of non-AML cells (GI50s = 1-10 µM); also inhibits the growth of DMS114 lung, HRA-19 colon, and RERF-LC-Sq1 lung cancer cells (GI50s = 16.6, 15.6, and 17.1 µM, respectively)
Catalog No. 50-294-1203