Learn More
Cayman Chemical ANTIBODY nI 9 5mg
SDP
Supplier:  Cayman Chemical 297335 MG
Encompass_Preferred
An inhibitor of ANO1 that blocks ATP-induced apical membrane chloride currents in ANO1-expressing FRT cells (IC50 = 77 nM); selective for ANO1 over ANO2 at 10 μM and CFTR and ENaC at 30 μM; inhibits ATP-induced quenching of a fluorescent intracellular halide sensor in PC3 prostate cancer cells and Capan-1 pancreatic carcinoma cells in a concentration-dependent manner
Catalog No. 50-298-9138