Learn More
Cayman Chemical ANTIBODY NIfenazon 500mg
SDP
Supplier:  Cayman Chemical 33330500 MG
Encompass_Preferred
A 20S proteasome activator; activates the chymotryptic-like peptidase activity of the yeast 20S proteasome in a cell-free assay at 0.5 µM; reduces Aβ42-induced decreases in mitochondrial activity and cell death in neuronally differentiated SH-SY5Y cells
Catalog No. 50-294-7179