Learn More
Cayman Chemical ANTIBODY nIracetam 10mg
SDP
Supplier:  Cayman Chemical 1775210 MG
Encompass_Preferred
A nootropic that acts as a dual allosteric modulator of AMPA and metabotropic glutamate receptors
Catalog No. 50-297-9801