Learn More
Cayman Chemical ANTIBODY NIthIamIde 5g
SDP
Supplier:  Cayman Chemical 360065 G
Encompass_Preferred
An antiprotozoal agent; active against G. intestinalis and T. vaginalis (IC50s = 0.49 and 0.022 µM, respectively); reduces mortality in a turkey model of histomoniasis at 0.05% in the diet
Catalog No. 50-291-2524