Learn More
Cayman Chemical ANTIBODY NItrocefIn 1mg
SDP
Supplier:  Cayman Chemical 154241 MG
Encompass_Preferred
A chromogenic cephalosporin substrate commonly used to detect β-lactamases in bacteria and other microbes
Catalog No. 50-296-9895