Learn More
Cayman Chemical ANTIBODY NIvalenol 1mg
SDP
Supplier:  Cayman Chemical 114381 MG
Encompass_Preferred
A trichothecene mycotoxin; lethal to mice (LD50 = 6.9 mg/kg); induces thymic, splenic, and Peyer's patch cell apoptosis in mice at 5, 10, and 15 mg/kg
Catalog No. 50-295-2420