Learn More
Cayman Chemical ANTIBODY NK 252 5mg
SDP
Supplier:  Cayman Chemical 191315 MG
Encompass_Preferred
A Nrf2 activator that interacts directly with the domain containing the Nrf2-binding site of Keap1; downregulates the expression of fibrogenic genes, producing antifibrotic effects in a rat model of non-alcoholic steatohepatitis
Catalog No. 50-298-1178