Learn More
Cayman Chemical ANTIBODY Nocodazole 10mg
SDP
Supplier:  Cayman Chemical 1385710 MG
Encompass_Preferred
An antimitotic agent; increases tubulin GTP hydrolysis and induces depolymerization; halts the cell cycle in the mitotic phase and synchronizes WI38 cells in the G2/M phase at 4 UG/ml; decreases neuronal mitochondrial transport and density in a transgenic zebrafish model with neurons expressing fluorescent proteins in mitochondrial and cell membranes at 400 nM
Catalog No. 50-295-5321