Learn More
Cayman Chemical ANTIBODY nonacIn 1mg
SDP
Supplier:  Cayman Chemical 224151 MG
Encompass_Preferred
A mitochondrial complex I inhibitor from A. muricata (IC50 = 54.8 nM); induces ATP depletion, tau pathologies, and dopaminergic cell death in rats (EC50s = 134, 44.1, and 60.8 nM, respectively); reduces cell survival (ED50 = 0.31 µM in MCF-7 cells); decreases MCF-7 xenograft tumor size in nude mice at a dose of 50 mg/kg per day
Catalog No. 50-298-6886