Learn More
Cayman Chemical ANTIBODY NS 1619 5mg
SDP
Supplier:  Cayman Chemical 188765 MG
Encompass_Preferred
A calcium-dependent activator of BKCa channels that is typically used at a concentration of 30 µM to hyperpolarize the membrane potential of single myocytes during studies of smooth muscle relaxation and vasodilation; cardioprotective against pulmonary hypertension as well as ischemia and reperfusion injury in various animal models
Catalog No. 50-298-4891