Learn More
Cayman Chemical ANTIBODY NSC 15520 25mg
SDP
Supplier:  Cayman Chemical 2602025 MG
Encompass_Preferred
A small molecule inhibitor of RPA70 dsDNA binding; selectively inhibits dsDNA over ssDNA binding to RPA70 from 25-400 µM; inhibits the protein-protein interaction between p53 or Rad9 and RPA70
Catalog No. 50-293-5853