Learn More
Cayman Chemical ANTIBODY NSC 5844 25mg
SDP
Supplier:  Cayman Chemical 2344325 MG
Encompass_Preferred
A bis-quinoline with diverse biological activities; inhibits the growth of P. falciparum D-6 and W-2 strains in vitro (IC50s = 17 and 27 nM, respectively); inhibits the growth of MDA-MB-468 and MCF-7 breast cancer cells (GI50s = 7.35 and 14.80 μM, respectively); inhibits 24% of BoNT/A LC metalloprotease activity at a concentration of 20 μM
Catalog No. 50-298-6753