Learn More
Cayman Chemical ANTIBODY NSC 617145 1mg
SDP
Supplier:  Cayman Chemical 219731 MG
Encompass_Preferred
An inhibitor of WRN helicase (IC50 = 230 nM); inhibits the ATPase, but not exonuclease, activity of WRN helicase in a concentration-dependent manner; inhibits growth of HeLa cells via formation of DSBs and induction of apoptosis in a WRN helicase-dependent manner
Catalog No. 50-298-7708