Learn More
Cayman Chemical ANTIBODY NSC 625987 5mg
SDP
Supplier:  Cayman Chemical 328435 MG
Encompass_Preferred
A Cdk4/cyclin D1 complex inhibitor (IC50 = 0.2 µM); selective for the Cdk4/cyclin D1 complex over Cdk2/cyclin A, Cdk2/cyclin E, and Cdk1/cyclin A complexes (IC50s = >100 µM for all)
Catalog No. 50-294-7886