Learn More
Cayman Chemical ANTIBODY NSC 756093 25mg
SDP
Supplier:  Cayman Chemical 3502325 MG
Encompass_Preferred
An inhibitor of the GBP1-Pim-1 protein-protein interaction; binds to GBP1-Pim-1 (Kd = 38 nM) and inhibits the interaction by 65% at 100 nM; enhances radiation-induced cytotoxicity in FaDu cells (IC50 = 496 nM); induces apoptosis and cell cycle arrest at the G2/M phase in FaDu cells
Catalog No. 50-291-3145