Learn More
Cayman Chemical ANTIBODY NU 6140 1mg
SDP
Supplier:  Cayman Chemical 172711 MG
Encompass_Preferred
A Cdk2 inhibitor (IC50 = 0.41 μM) that demonstrates 10- to 36-fold selectivity against Cdk2-cyclin A compared to Cdk1-cyclin B, Cdk4-cyclin D, Cdk5-p25, or Cdk7-cyclin H; induces cell-cycle arrest at the G2-M phase
Catalog No. 50-298-0542