Learn More
Cayman Chemical ANTIBODY NulscrIpt 10mg
SDP
Supplier:  Cayman Chemical 1643310 MG
Encompass_Preferred
An analog of scriptaid that lacks scriptaid’s HDAC inhibitory effects; used as a negative control in experiments involving scriptaid
Catalog No. 50-297-0576