Learn More
Cayman Chemical ANTIBODY NVP-TAE226 1mg
SDP
Supplier:  Cayman Chemical 176851 MG
Encompass_Preferred
A FAK inhibitor (IC50 = 5.5 nM) that also inhibits IGF-1R (IC50 = 0.16 µM); inhibits glioma and ovarian tumor growth in in vivo tumor models
Catalog No. 50-298-0170